AC5388701412 Accela ChemBio Inc.

Ethyl 2-(2-Methoxy-5-pyrimidinyl)-2-oxoacetate

  • MDL number MFCD20134207
  • Cas Number : 1346597-52-1
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016040&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016040.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart