AC5388701450 Accela ChemBio Inc.

6-Methyl-5-cyanouracil

  • MDL number MFCD07355755
  • Cas Number : 5900-40-3
  • Molecular Weight : 151.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017223&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017223.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart