AC5388701458 Accela ChemBio Inc.

4-(4-Bromophenyl)tetrahydropyran-4-ol

  • MDL number MFCD11849469
  • Cas Number : 165119-46-0
  • Molecular Weight : 257.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017428&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017428.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart