AC5388701501 Accela ChemBio Inc.

Ethyl 4-(Trifluoromethyl)thiazole-2-carboxylate

  • MDL number MFCD08669898
  • Cas Number : 79247-86-2
  • Molecular Weight : 225.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018404&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018404.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart