AC5388701513 Accela ChemBio Inc.

6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine

  • MDL number MFCD08447399
  • Cas Number : 53493-80-4
  • Molecular Weight : 121.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid, or colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018652&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018652.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart