AC5388701519 Accela ChemBio Inc.

4-Bromo-2,6-dinitroaniline

  • MDL number MFCD00955747
  • Cas Number : 62554-90-9
  • Molecular Weight : 262.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2,6-dinitroaniline
Inchi Code 1S/C6H4BrN3O4/c7-3-1-4(9(11)12)6(8)5(2-3)10(13)14/h1-2H,8H2
Inchi Key IIUAAKYBFINVAD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018666&attname=GoodsCode

Safety Information

signalword Warning
MSDS http://pdf.accelachem.com/cn/msds/SY018666.pdf
hcodes GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart