AC5388701521 Accela ChemBio Inc.

1-Boc-3-ethylpiperidine-3-carboxylic Acid

  • MDL number MFCD12406586
  • Cas Number : 887591-65-3
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tert-butoxycarbonyl)-3-ethyl-3-piperidinecarboxylic acid
Inchi Code 1S/C13H23NO4/c1-5-13(10(15)16)7-6-8-14(9-13)11(17)18-12(2,3)4/h5-9H2,1-4H3,(H,15,16)
Inchi Key NKZAZUYDWWUQBL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018685&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018685.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart