AC5388701522 Accela ChemBio Inc.

4-Bromo-2-[(methylamino)methyl]phenol

  • MDL number MFCD09965827
  • Cas Number : 157729-23-2
  • Molecular Weight : 216.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018706&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018706.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart