AC5388701524 Accela ChemBio Inc.

Isopropylsulfamoyl Chloride

  • MDL number MFCD09753748
  • Cas Number : 26118-67-2
  • Molecular Weight : 157.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to brown liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018712&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018712.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart