AC5388701525 Accela ChemBio Inc.

Isobutylsulfamoyl Chloride

  • MDL number MFCD11520374
  • Cas Number : 26118-68-3
  • Molecular Weight : 171.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018714&attname=GoodsCode

Safety Information

signalword Danger
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018714.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart