AC5388701544 Accela ChemBio Inc.

1-Benzyl-2-[3-(1-benzyl-1,3-dimethyl-1H-benzo[e]indol-2(3H)-ylidene)-1-propen-1-yl]-1,3-dimethyl-1H-benzo[e]indol-3-ium Hexafluorophosphate

  • MDL number MFCD18071595
  • Cas Number : 581092-53-7
  • Molecular Weight : 754.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019568&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019568.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart