AC5388701549 Accela ChemBio Inc.

Benzyl N-Benzylpyrrolidine-3-carboxylate

  • MDL number MFCD13190633
  • Cas Number : 570423-97-1
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019796&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019796.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart