AC5388701570 Accela ChemBio Inc.

N-Hydroxyoctanamide

  • MDL number MFCD00143918
  • Cas Number : 7377-03-9
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020160&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY020160.pdf
hstatements H315-H318-H335
Availability details for
Quantity
  • Availability
  • Add to Cart