AC5388701586 Accela ChemBio Inc.

1-(4-Bromo-2-fluorophenyl)-3-chloro-1-propanone

  • MDL number MFCD22047095
  • Cas Number : 395639-63-1
  • Molecular Weight : 265.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020444&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020444.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart