AC5388701656 Accela ChemBio Inc.

Dimethyl 2-Cyanosuccinate

  • MDL number MFCD01760090
  • Cas Number : 6283-71-2
  • Molecular Weight : 171.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021147&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021147.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart