AC5388701659 Accela ChemBio Inc.

7-Methoxyquinoline Hydrochloride

  • MDL number MFCD22683321
  • Cas Number : 1418117-82-4
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021167&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021167.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart