AC5388701669 Accela ChemBio Inc.

1-Boc-3-(3,4-dichlorophenyl)piperazine

  • MDL number MFCD04038531
  • Cas Number : 185110-16-1
  • Molecular Weight : 331.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021226&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021226.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart