AC5388701704 Accela ChemBio Inc.

7-Aminopyrazolo[1,5-a]pyrimidine-3,6-dicarbonitrile

  • MDL number MFCD01812292
  • Cas Number : 119270-27-8
  • Molecular Weight : 184.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-aminopyrazolo[1,5-a]pyrimidine-3,6-dicarbonitrile
Inchi Code 1S/C8H4N6/c9-1-5-3-12-8-6(2-10)4-13-14(8)7(5)11/h3-4H,11H2
Inchi Key NXEKAPRNZKNMOD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021430&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021430.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart