AC5388701707 Accela ChemBio Inc.

4-Fluoro-1H-indazole-5-carbonitrile

  • MDL number MFCD09743858
  • Cas Number : 473416-81-8
  • Molecular Weight : 161.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021437&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021437.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart