AC5388701708 Accela ChemBio Inc.

4-Hydroxythieno[2,3-b]pyridine-5-carbonitrile

  • MDL number MFCD12755339
  • Cas Number : 63873-60-9
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021440&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021440.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart