AC5388701710 Accela ChemBio Inc.

2-(Hydroxymethyl)-5-nitrothiophene

  • MDL number MFCD06202767
  • Cas Number : 20898-85-5
  • Molecular Weight : 159.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-nitro-2-thienyl)methanol
Inchi Code 1S/C5H5NO3S/c7-3-4-1-2-5(10-4)6(8)9/h1-2,7H,3H2
Inchi Key QYNOWSBTIMNUDS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form Light yellow to orange solid or liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021452&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021452.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart