AC5388701721 Accela ChemBio Inc.

N-Acetyl-(2R)-bornane-10,2-sultam

  • MDL number MFCD00269649
  • Cas Number : 141993-16-0
  • Molecular Weight : 257.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021531&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P302+P352-P280-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY021531.pdf
hstatements H319-H315-H335
Availability details for
Quantity
  • Availability
  • Add to Cart