AC5388701723 Accela ChemBio Inc.

N-Propionyl-(2R)-bornane-10,2-sultam

  • MDL number MFCD00269689
  • Cas Number : 125664-95-1
  • Molecular Weight : 271.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021533&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021533.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart