AC5388701724 Accela ChemBio Inc.

N-Propionyl-(2S)-bornane-10,2-sultam

  • MDL number MFCD00269690
  • Cas Number : 128947-19-3
  • Molecular Weight : 271.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021534&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021534.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart