AC5388701725 Accela ChemBio Inc.

3-(Boc-aminomethyl)indoline

  • MDL number MFCD09971405
  • Cas Number : 1000932-73-9
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021536&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021536.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart