AC5388701746 Accela ChemBio Inc.

Methyl 5-Formyl-2-thiophenecarboxylate

  • MDL number MFCD01859942
  • Cas Number : 67808-64-4
  • Molecular Weight : 170.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021708&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021708.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart