AC5388701748 Accela ChemBio Inc.

1-Boc-4-piperidinesulfonyl Chloride

  • MDL number MFCD01861750
  • Cas Number : 782501-25-1
  • Molecular Weight : 283.77222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021733&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280-P301+P330+P331-P310-P305+P351+P338-P303+P361+P353-P402+P404
hcodes GHS05
MSDS http://pdf.accelachem.com/cn/msds/SY021733.pdf
hstatements H314-EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart