AC5388701749 Accela ChemBio Inc.

5-Bromo-3-furancarboxylic Acid

  • MDL number MFCD08669547
  • Cas Number : 58832-36-3
  • Molecular Weight : 190.97952

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021737&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021737.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart