AC5388701751 Accela ChemBio Inc.

2-[(Methylamino)methyl]benzyl Alcohol

  • MDL number MFCD09035678
  • Cas Number : 57685-24-2
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021761&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY021761.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart