AC5388701752 Accela ChemBio Inc.

2,4-Difluoro-3-formylbenzonitrile

  • MDL number MFCD09907858
  • Cas Number : 149489-14-5
  • Molecular Weight : 167.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021763&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021763.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart