AC5388701754 Accela ChemBio Inc.

5-Aminothieno[2,3-d]pyrimidine-6-carboxylic Acid

  • MDL number MFCD09701987
  • Cas Number : 59488-81-2
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021773&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021773.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart