AC5388701788 Accela ChemBio Inc.

3-Bromo-4-(chloromethyl)pyridine Hydrochloride

  • MDL number MFCD22682795
  • Cas Number : 1418117-80-2
  • Molecular Weight : 242.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021915&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021915.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart