AC5388701798 Accela ChemBio Inc.

6-Amino-1H-indazol-3(2H)-one Hydrochloride

  • MDL number MFCD16037326
  • Cas Number : 220991-84-4
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021946&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021946.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart