AC5388701806 Accela ChemBio Inc.

4-Methoxy-2-(trifluoromethyl)benzamidine Hydrochloride

  • MDL number MFCD22574993
  • Cas Number : 1408058-12-7
  • Molecular Weight : 254.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021959&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021959.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart