AC5388701817 Accela ChemBio Inc.

4-(4-Methylphenyl)-2-butanone

  • MDL number MFCD00156655
  • Cas Number : 7774-79-0
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022010&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022010.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart