AC5388701828 Accela ChemBio Inc.

Bis(4-biphenylyl)amine

  • MDL number MFCD08276279
  • Cas Number : 102113-98-4
  • Molecular Weight : 321.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022030&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P350-P305+P351+P338-P332+P313-P337+P313-P362
hcodes GHS07,
MSDS http://pdf.accelachem.com/cn/msds/SY022030.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart