AC5388701839 Accela ChemBio Inc.

Isobutyl 2-(Methylamino)benzoate

  • MDL number MFCD14586757
  • Cas Number : 65505-24-0
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022057&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022057.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart