AC5388701845 Accela ChemBio Inc.

Methyl 5-Oxo-5-(2-thienyl)pentanoate

  • MDL number MFCD10567619
  • Cas Number : 18760-47-9
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022076&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022076.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart