AC5388701848 Accela ChemBio Inc.

tert-Butylhydroquinone Diacetate

  • MDL number MFCD00078251
  • Cas Number : 7507-48-4
  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022082&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022082.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart