AC5388701855 Accela ChemBio Inc.

(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine Dihydrochloride

  • MDL number MFCD09749879
  • Cas Number : 70708-33-7
  • Molecular Weight : 215.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022102&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P302+P352-P304+P340-P261-P312
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY022102.pdf
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart