AC5388701876 Accela ChemBio Inc.

3-(3-Pyridyl)-2-propen-1-ol

  • MDL number MFCD03425627
  • Cas Number : 69963-46-8
  • Molecular Weight : 135.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022168&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY022168.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart