AC5388701877 Accela ChemBio Inc.

3-(2-Pyridyl)-2-propen-1-ol

  • MDL number MFCD18384384
  • Cas Number : 131610-03-2
  • Molecular Weight : 135.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022169&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY022169.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart