AC5388701878 Accela ChemBio Inc.

2-Bromo-5-(trifluoromethyl)phenylhydrazine Hydrochloride

  • MDL number MFCD03094412
  • Cas Number : 529512-78-5
  • Molecular Weight : 291.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022172&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY022172.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart