AC5388701880 Accela ChemBio Inc.

3-Hydrazinobenzoic Acid Hydrochloride

  • MDL number MFCD11226177
  • Cas Number : 87565-98-8
  • Molecular Weight : 188.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022176&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY022176.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart