AC5388701886 Accela ChemBio Inc.

1,2,3,4-Tetrahydrophthalazine Dihydrochloride

  • MDL number MFCD22574996
  • Cas Number : 86437-12-9
  • Molecular Weight : 207.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022231&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022231.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart