AC5388701889 Accela ChemBio Inc.

3-(Aminomethyl)indoline Dihydrochloride

  • MDL number MFCD22574997
  • Cas Number : 1408058-15-0
  • Molecular Weight : 221.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022297&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022297.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart