AC5388701890 Accela ChemBio Inc.

DL-Isoleucine

  • MDL number MFCD00004268
  • Cas Number : 443-79-8
  • Molecular Weight : 131.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022340&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022340.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart