AC5388701894 Accela ChemBio Inc.

2-Amino-2-(2-pyridyl)acetic Acid

  • MDL number MFCD05225556
  • Cas Number : 62451-88-1
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022365&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022365.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart