AC5388701895 Accela ChemBio Inc.

1-Boc-3-bromo-5-nitro-1H-indazole

  • MDL number MFCD11617131
  • Cas Number : 473416-22-7
  • Molecular Weight : 342.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022369&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022369.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart