AC5388701896 Accela ChemBio Inc.

N-Boc-N-methyl-3-chloro-1-propanamine

  • MDL number MFCD09031544
  • Cas Number : 114326-14-6
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022383&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022383.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart